5-Bromopyrimidine ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
B2229-25G B2229-5G
TCIAB2229-25GEA 214 EUR
TCIAB2229-25G TCIAB2229-5G
5-Bromopyrimidine ≥98.0% (by GC)
5-Bromopyrimidine
Formule: C₄H₃BrN₂
Kookpunt: 168°C
Smeltpunt: 75…76 °C
MDL Nummer: MFCD00006117
CAS nummer: 4595-59-9
EINECS: 224-992-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8738b399a64a6c604ca27362b0873becc7a8f41&vid=d8738b399a64a6c604ca27362b0873becc7a8f41&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 72.0 to 76.0 deg-C