Indole-5-carbonitrile ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 1H-Indole-5-carbonitrile, 5-Cyanoindole, 5-Cyano-1H-indole

C1491-1G C1491-25G C1491-5G
TCIAC1491-1GEA 31.9 EUR
TCIAC1491-1G TCIAC1491-25G TCIAC1491-5G
Indole-5-carbonitrile ≥98.0% (by GC)
Indole-5-carbonitrile
Formule: C₉H₆N₂
Smeltpunt: 106…108 °C
MDL Nummer: MFCD00005669
CAS nummer: 15861-24-2
EINECS: 239-986-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ac0ffd70e1f00e70589b1ceca8207b8b5c38c36&vid=3ac0ffd70e1f00e70589b1ceca8207b8b5c38c36&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 105.0 to 108.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR