4-Pentyn-1-ol ≥96.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
P0817-25ML
P0817-5ML
TCIAP0817-25MLEA
137
EUR
TCIAP0817-25ML
TCIAP0817-5ML
4-Pentyn-1-ol ≥96.0% (by GC)
4-Pentyn-1-ol
Formule:
C₅H₈O Kookpunt: 155°C Dichtheid: 0,91 g/cm³ (20 °C) Vlampunt: 62°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00002974 CAS nummer: 5390-04-5 EINECS: 226-383-6 |
|
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...