6-Methoxyindole ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 6-Methoxy-1H-indole, 1H-Indol-6-yl methyl ether, Indol-6-yl methyl ether

M1214-1G M1214-100MG
TCIAM1214-1GEA 118 EUR
TCIAM1214-1G TCIAM1214-100MG
6-Methoxyindole ≥98.0% (by GC)
6-Methoxyindole
Formule: C₉H₉NO
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00022780
CAS nummer: 3189-13-7
EINECS: 221-689-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=434bbdb853ddec1d2288f61d211531fb4daf8502&vid=434bbdb853ddec1d2288f61d211531fb4daf8502&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Gray to Red powder to crystal
Purity (GC) min. 98.0 %
Melting point 89.0 to 94.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR