Broomazijnzuur ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

B0531-500G B0531-25G
TCIAB0531-500GEA 104 EUR
TCIAB0531-500G TCIAB0531-25G
Broomazijnzuur ≥98.0% (by titrimetric analysis)
Broomazijnzuur
Formule: BrCH₂COOH
Kookpunt: 206 °C (1013 hPa)
Smeltpunt: 46…49 °C
Dichtheid: 1,9335 g/cm³ (20 °C)
Vlampunt: ≥ 112 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002678
CAS nummer: 79-08-3
EINECS: 201-175-8
UN: 3425
ADR: 8,II
Merck Index: 14,01400
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=908eb5d562f2e7f48e399a33b5eef6dd738e807e&vid=908eb5d562f2e7f48e399a33b5eef6dd738e807e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to lump
Purity (Neutralization titration) min. 98.0 %
Melting point 46.0 to 50.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR