Methyl butyrate ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Butyric acid methyl ester, Butanoic acid methyl ester

B0763-25ML B0763-500ML
TCIAB0763-25MLEA 17.6 EUR
TCIAB0763-25ML TCIAB0763-500ML
Methyl butyrate ≥99.0% (by GC)
Methyl butyrate
Formule: C₅H₁₀O₂
Kookpunt: 101°C
Smeltpunt: –84 °C
Dichtheid: 0,8984 g/cm³ (20 °C)
Vlampunt: 13°C
MDL Nummer: MFCD00009391
CAS nummer: 623-42-7
EINECS: 210-792-1
UN: 1237
ADR: 3,II
Merck Index: 14,06035
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc840a078ae10e94ba9712ab14e81d06bc98756f&vid=fc840a078ae10e94ba9712ab14e81d06bc98756f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR