Dimethyl maleate ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Dimethyl Maleate, Maleic acid dimethyl ester
M0012-25ML M0012-500ML
TCIAM0012-25MLEA 18.7 EUR
TCIAM0012-25ML TCIAM0012-500ML
Dimethyl maleate ≥97.0% (by GC)
Dimethyl maleate
Formule: C₆H₈O₄
Kookpunt: 205°C
Smeltpunt: -19°C
Dichtheid: 1,153 g/cm³ (20 °C)
Vlampunt: 103°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008459
CAS nummer: 624-48-6
EINECS: 210-848-5
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c677d2d94d8896a9307229978d76314e6531d906&vid=c677d2d94d8896a9307229978d76314e6531d906&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %