Dodecyltriethoxysilane ≥95.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Triethoxy(dodecyl)silane
D1510-10ML
D1510-25ML
TCIAD1510-10MLEA
40.7
EUR
TCIAD1510-10ML
TCIAD1510-25ML
Dodecyltriethoxysilane ≥95.0% (by GC)
Dodecyltriethoxysilane
Formule:
C₁₈H₄₀O₃Si MW: 332.6 g/mol Kookpunt: 129°C |
MDL Nummer:
MFCD00048437 CAS nummer: 18536-91-9 EINECS: 242-409-9 |
|
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...