Ethanesulphonyl chloride ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
E0035-25G E0035-250G
TCIAE0035-25GEA 19.8 EUR
TCIAE0035-25G TCIAE0035-250G
Ethanesulphonyl chloride ≥98.0% (by titrimetric analysis)
Ethanesulphonyl chloride
Formule: C₂H₅ClO₂S
Kookpunt: 171°C
Smeltpunt: –68 °C
Dichtheid: 1,36 g/cm³ (20 °C)
Vlampunt: 88°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007460
CAS nummer: 594-44-5
EINECS: 209-842-5
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd8959a1ab554fdda35e91dd29ae46b54e95849e&vid=bd8959a1ab554fdda35e91dd29ae46b54e95849e&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (Argentmetric Titration) min. 98.0 %