Heptafluorobutyric acid ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Perfluorobutyric acid, HFBA, Heptafluorobutanoic acid
H0024-100G
H0024-25G
TCIAH0024-100GEA
294
EUR
TCIAH0024-100G
TCIAH0024-25G
Heptafluorobutyric acid ≥98.0% (by GC, titration analysis)
Heptafluorobutyric acid
Formule:
C₄HF₇O₂ MW: 214.04 g/mol Kookpunt: 120°C Smeltpunt: –20°C Dichtheid: 1.764 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00004171 CAS nummer: 375-22-4 EINECS: 206-786-3 UN: 3265 ADR: 8 II |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...