Cetrimonium bromide ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

H0081-25G H0081-500G H0081-100G
TCIAH0081-25GEA 17.6 EUR
TCIAH0081-25G TCIAH0081-500G TCIAH0081-100G
Cetrimonium bromide ≥98.0% (by titrimetric analysis)
Cetrimonium bromide
Formule: CH₃(CH₂)₁₅N(Br)(CH₃)₃
Smeltpunt: 243°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011772
CAS nummer: 57-09-0
EINECS: 200-311-3
UN: 3077
ADR: 9,III
Merck Index: 14,02025
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8793e318ade2880426390ae764ade9cb3e089bd3&vid=8793e318ade2880426390ae764ade9cb3e089bd3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR