Ethyl stearate ≥96.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Stearic acid ethyl ester, Ethyl octadecanoate
S0079-25G S0079-250G
TCIAS0079-25GEA 26.4 EUR
TCIAS0079-25G TCIAS0079-250G
Ethyl stearate ≥96.0% (by GC)
Ethyl stearate
Formule: C₂₀H₄₀O₂
Kookpunt: 199°C
Smeltpunt: 31 °C
Dichtheid: 0,8973 g/cm³ (25 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00009006
CAS nummer: 111-61-5
EINECS: 203-887-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=850e31ba455f853b0fb68eab8e2b0f97adc3baee&vid=850e31ba455f853b0fb68eab8e2b0f97adc3baee&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 96.0 %