Triethylboraat ≥97.0% (by titrimetric analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
B0520-25ML B0520-500ML B0520-100ML
TCIAB0520-25MLEA 17.6 EUR
TCIAB0520-25ML TCIAB0520-500ML TCIAB0520-100ML
Triethylboraat ≥97.0% (by titrimetric analysis)
Triethylboraat
Formule: C₆H₁₅BO₃
Kookpunt: 118°C
Smeltpunt: –85°C
Dichtheid: 0,858 g/cm³ (20 °C)
Vlampunt: 11°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012846
CAS nummer: 150-46-9
EINECS: 205-760-9
UN: 1176
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7b6e5251e8559cc3d938d80d701ef62910f2a38&vid=c7b6e5251e8559cc3d938d80d701ef62910f2a38&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (Neutralization titration) min. 97.0 %