Pentafluoropropionic acid ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Perfluoropropionic acid, Pentafluoropropanoic acid, Perfluoropropanoic acid, 2,2,3,3,3-Pentafluoropropionic acid

P1125-25G P1125-100G
TCIAP1125-25GEA 117 EUR
TCIAP1125-25G TCIAP1125-100G
Pentafluoropropionic acid ≥98.0% (by GC, titration analysis)
Pentafluoropropionic acid
Formule: C₃HF₅O₂
MW: 164.03 g/mol
Kookpunt: 98°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004170
CAS nummer: 422-64-0
EINECS: 207-021-6
UN: 3265
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=395be14e210c15ac9dfcb0a8b47ddab056045bc6&vid=395be14e210c15ac9dfcb0a8b47ddab056045bc6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR