Polyethylene glycol 400
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: PEG 400
N0443-25G N0443-500G
TCIAN0443-25GEA 17.6 EUR
TCIAN0443-25G TCIAN0443-500G
Polyethylene glycol 400
Polyethylene glycol 400
Formule: H(OCH₂CH₂)nOH
MW: 400 g/mol
Vlampunt: 224°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00081839
CAS nummer: 25322-68-3
EINECS: 500-038-2
Merck Index: 14,07568
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Average molecular weight 380 to 420