Propionamide ≥98.0% (by GC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

P0499-25G P0499-500G
TCIAP0499-25GEA 73.7 EUR
TCIAP0499-25G TCIAP0499-500G
Propionamide ≥98.0% (by GC, total nitrogen)
Propionamide
Formule: C₃H₇NO
Kookpunt: 213°C
Smeltpunt: 79 °C
Dichtheid: 1,1 g/cm³ (20 °C)
MDL Nummer: MFCD00008039
CAS nummer: 79-05-0
EINECS: 201-172-1
Merck Index: 14,07824
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3efef2ceb24445a039073ff27b2510742068d86f&vid=3efef2ceb24445a039073ff27b2510742068d86f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 77.0 to 82.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR