Tetradecylcyclohexane ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

T0873-25G T0873-5G
TCIAT0873-25GEA 900 EUR
TCIAT0873-25G TCIAT0873-5G
Tetradecylcyclohexane ≥98.0% (by GC)
Tetradecylcyclohexane
Formule: C₂₀H₄₀
Kookpunt: 360 °C (1013 hPa)
Smeltpunt: 42 °C
Dichtheid: 0,8254 g/cm³ (20 °C)
MDL Nummer: MFCD00058951
CAS nummer: 1795-18-2
EINECS: 217-274-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10ef0b957a7d61aace986e1cf3a7f67f2bb7797b&vid=10ef0b957a7d61aace986e1cf3a7f67f2bb7797b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 98.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR