Methylthiocyanaat ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
T0201-25ML T0201-500ML
TCIAT0201-25MLEA 38.5 EUR
TCIAT0201-25ML TCIAT0201-500ML
Methylthiocyanaat ≥98.0% (by GC)
Methylthiocyanaat
Formule: C₂H₃NS
Kookpunt: 130°C
Smeltpunt: –5 °C
Dichtheid: 1,07 g/cm³ (20 °C)
Vlampunt: 38°C
MDL Nummer: MFCD00001830
CAS nummer: 556-64-9
EINECS: 209-134-6
UN: 1992
ADR: 3,III
Merck Index: 14,06127
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6edfda71961d19faefd697571d0a3d882abfac09&vid=6edfda71961d19faefd697571d0a3d882abfac09&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %