Triethylsilane ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
T0662-250ML
T0662-25ML
TCIAT0662-250MLEA
138
EUR
TCIAT0662-250ML
TCIAT0662-25ML
Triethylsilane ≥98.0% (by GC)
Triethylsilane
Formule:
C₆H₁₆Si Kookpunt: 108°C Dichtheid: 0,73 g/cm³ (20 °C) Vlampunt: –3°C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00009018 CAS nummer: 617-86-7 EINECS: 210-535-3 UN: 1993 ADR: 3,II |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...