(2R,3R)-(-)-2,3-Butanediol ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B1161-1G B1161-5G
TCIAB1161-1GEA 56.1 EUR
TCIAB1161-1G TCIAB1161-5G
(2R,3R)-(-)-2,3-Butanediol ≥97.0% (by GC)
(2R,3R)-(-)-2,3-Butanediol
Formule: C₄H₁₀O₂
Kookpunt: 80°C
Smeltpunt: 16 °C
Dichtheid: 0,98 g/cm³ (20 °C)
Vlampunt: 85°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064267
CAS nummer: 24347-58-8
EINECS: 246-186-9
Merck Index: 13,01567
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=deed47ce80da158ba72a1104db79df4cef9bb4ac&vid=deed47ce80da158ba72a1104db79df4cef9bb4ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 97.0 %
Optical purity (GC) min. 98.0 ee%

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR