(R)-(-)-2-Amino-3-methyl-1-butanol ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
D(-)-2-Amino-3-methyl-1-butanol, (R)-(-)-2-Amino-3-methylbutanol, D-Valinol, D(-)-Valinol
V0077-1G
V0077-5G
TCIAV0077-1GEA
26.4
EUR
TCIAV0077-1G
TCIAV0077-5G
(R)-(-)-2-Amino-3-methyl-1-butanol ≥98.0% (by GC, titration analysis)
(R)-(-)-2-Amino-3-methyl-1-butanol
Formule:
C₅H₁₃NO MW: 103.16 g/mol |
MDL Nummer:
MFCD00064297 CAS nummer: 4276-09-9 |
|
Specification Test Results
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Optical purity (GC) | min. 98.0 ee% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...