4'-Chloroacetanilide ≥98.0% (by GC, total nitrogen)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
C0089-25G
C0089-500G
TCIAC0089-25GEA
50.6
EUR
TCIAC0089-25G
TCIAC0089-500G
4'-Chloroacetanilide ≥98.0% (by GC, total nitrogen)
4'-Chloroacetanilide
Formule:
C₈H₈ClNO Kookpunt: 333 °C (1013 hPa) Smeltpunt: 179…180 °C Dichtheid: 1,385 g/cm³ (22 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00000612 CAS nummer: 539-03-7 EINECS: 208-707-8 Merck Index: 14,02111 |
|
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 176.0 to 182.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...