Ethyl pyruvate ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Ethyl 2-oxopropanoate, 2-Oxopropionic acid ethyl ester, α-Ketopropionic acid ethyl ester, Pyruvic acid ethyl ester, 2-Ketopropionic acid ethyl ester

P0891-25ML P0891-500ML P0891-100ML
TCIAP0891-25MLEA 17.6 EUR
TCIAP0891-25ML TCIAP0891-500ML TCIAP0891-100ML
Ethyl pyruvate ≥97.0% (by GC)
Ethyl pyruvate
Formule: C₅H₈O₃
MW: 116.12 g/mol
Kookpunt: 142°C
Smeltpunt: -50
Vlampunt: 51°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00009123
CAS nummer: 617-35-6
EINECS: 210-511-2
UN: 3272
ADR: 3 III
Merck Index: 14,08021
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a81401d537521c3d159ae1ae85aafe70deb332bb&vid=a81401d537521c3d159ae1ae85aafe70deb332bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR