Ethyl acetoacetate ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
A0649-25G A0649-500G
TCIAA0649-25GEA 17.6 EUR
TCIAA0649-25G TCIAA0649-500G
Ethyl acetoacetate ≥98.0% (by GC)
Ethyl acetoacetate
Formule: C₆H₁₀O₃
Kookpunt: 184°C
Smeltpunt: –45 °C
Dichtheid: 1,0188 g/cm³ (20 °C)
Vlampunt: 75°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00009199
CAS nummer: 141-97-9
EINECS: 205-516-1
Merck Index: 14,03758
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44cb704dc6f8ecaaee0805da8c4000670b3bee73&vid=44cb704dc6f8ecaaee0805da8c4000670b3bee73&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %