Hexanoyl chloride ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
H0117-25G
H0117-500G
H0117-100G
TCIAH0117-25GEA
17.6
EUR
TCIAH0117-25G
TCIAH0117-500G
TCIAH0117-100G
Hexanoyl chloride ≥98.0% (by GC, titration analysis)
Hexanoyl chloride
Formule:
C₆H₁₁ClO MW: 134.61 g/mol Kookpunt: 152 Smeltpunt: -87 Vlampunt: 50°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00000760 CAS nummer: 142-61-0 EINECS: 205-549-1 UN: 2920 ADR: 8 II Merck Index: 14,01763 |
|
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...