4'-Nitroacetanilide ≥99.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
N-(4-Nitrophenyl)acetamide
N0108-25G
N0108-500G
TCIAN0108-25GEA
99
EUR
TCIAN0108-25G
TCIAN0108-500G
4'-Nitroacetanilide ≥99.0% (by GC)
4'-Nitroacetanilide
Formule:
C₈H₈N₂O₃ Kookpunt: 409 °C (1013 hPa) Smeltpunt: 215 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00007303 CAS nummer: 104-04-1 EINECS: 203-169-0 Merck Index: 14,06581 |
|
Specification Test Results
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 213.0 to 217.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...