Sebacoyl dichloride ≥95.0% (by GC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Sebacoyl chloride
S0030-25ML S0030-500ML S0030-100ML
TCIAS0030-25MLEA 28.6 EUR
TCIAS0030-25ML TCIAS0030-500ML TCIAS0030-100ML
Sebacoyl dichloride ≥95.0% (by GC, titration analysis)
Sebacoyl dichloride
Formule: C₁₀H₁₆Cl₂O₂
Kookpunt: 167°C
Smeltpunt: –3°C
Dichtheid: 1,12 g/cm³ (20 °C)
Vlampunt: 164°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00000770
CAS nummer: 111-19-3
EINECS: 203-843-4
UN: 2922
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0e0bf5b08fc9d6106953f7b191de31c4c8d2fad&vid=c0e0bf5b08fc9d6106953f7b191de31c4c8d2fad&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Brown to Dark purple clear liquid
Purity (GC) min. 95.0 %
Purity (Argentmetric Titration) min. 95.0 %