N-Lauroylsarcosine ≥90.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
L0151-25G
L0151-500G
L0151-100G
TCIAL0151-25GEA
17.6
EUR
TCIAL0151-25G
TCIAL0151-500G
TCIAL0151-100G
N-Lauroylsarcosine ≥90.0% (by GC)
N-Lauroylsarcosine
Formule:
H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COOH |
MDL Nummer:
MFCD00021749 CAS nummer: 97-78-9 EINECS: 202-608-3 UN: 2811 ADR: 6.1,II |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 90.0 % |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...