2-Aminomethyldimethylamine ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
N,N-Dimethylethylenediamine, asym. Dimethylethylendiamine, unsym-Dimethylethylenediamine, 2-(Dimethylamino)ethylamine, 1-Amino-2-dimethylaminoethane, 2-Dimethylaminoethylamine, 2-Aminoethyldimethylamine
D0719-25ML
D0719-500ML
D0719-100ML
TCIAD0719-25MLEA
27.5
EUR
TCIAD0719-25ML
TCIAD0719-500ML
TCIAD0719-100ML
2-Aminomethyldimethylamine ≥98.0% (by GC, titration analysis)
2-Aminomethyldimethylamine
Formule:
C₄H₁₂N₂ Kookpunt: 107°C Smeltpunt: –70 °C Dichtheid: 0,804 g/cm³ (20 °C) Vlampunt: 18°C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00008175 CAS nummer: 108-00-9 EINECS: 203-541-2 UN: 2733 ADR: 3,II |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...