2-Dimethylaminoethanol ≥99.0% (by GC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: DMAE
D0649-25ML D0649-500ML
TCIAD0649-25MLEA 18.7 EUR
TCIAD0649-25ML TCIAD0649-500ML
2-Dimethylaminoethanol ≥99.0% (by GC, titration analysis)
2-Dimethylaminoethanol
Formule: C₄H₁₁NO
MW: 89.14 g/mol
Kookpunt: 134°C
Smeltpunt: -59
Vlampunt: 40°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002846
CAS nummer: 108-01-0
EINECS: 203-542-8
UN: 2051
ADR: 8 II
Merck Index: 14,02843
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af2e6bda25e3edf4c98de4bd4ef36b452c1a327e&vid=af2e6bda25e3edf4c98de4bd4ef36b452c1a327e&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 99.0 %