Aluminiumisopropylaat ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
A0246-100G A0246-500G
TCIAA0246-100GEA 18.7 EUR
TCIAA0246-100G TCIAA0246-500G
Aluminiumisopropylaat ≥98.0% (by titrimetric analysis)
Aluminiumisopropylaat
Formule: C₉H₂₁AlO₃
Kookpunt: 135 °C (13 hPa)
Smeltpunt: 119 °C
Dichtheid: 0,9771 g/cm³ (20 °C)
MDL Nummer: MFCD00008870
CAS nummer: 555-31-7
EINECS: 209-090-8
UN: 1325
ADR: 4.1,II
Merck Index: 14,00347
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=550038a2b7065ce12699650703bd3169a048f5aa&vid=550038a2b7065ce12699650703bd3169a048f5aa&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to lump
Purity (Chelometric Titration) min. 98.0 %
Melting point 132.0 to 140.0 deg-C
Specificaties
Environmentally Preferable