Potassium methanesulphonate ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Potassiummethanesulphonate, Methanesulphonic acid potassium salt
M0550-100G
M0550-25G
TCIAM0550-100GEA
157
EUR
TCIAM0550-100G
TCIAM0550-25G
Potassium methanesulphonate ≥98.0% (by titrimetric analysis)
Potassium methanesulphonate
Formule:
CH₃KO₃S MW: 134.2 g/mol |
MDL Nummer:
MFCD00070544 CAS nummer: 2386-56-3 EINECS: 219-202-7 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Ion exchange titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Drying loss | max. 1.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...