Pivalic anhydride ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
P1414-25ML P1414-250ML
TCIAP1414-25MLEA 33 EUR
TCIAP1414-25ML TCIAP1414-250ML
Pivalic anhydride ≥98.0% (by GC)
Pivalic anhydride
Formule: C₁₀H₁₈O₃
Kookpunt: 193 °C (1013 hPa)
Dichtheid: 0,91 g/cm³ (20 °C)
Vlampunt: 57°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008842
CAS nummer: 1538-75-6
EINECS: 216-263-1
UN: 2920
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f90ab3baa6f4edc490da29b6673f25d2fa54d2e&vid=3f90ab3baa6f4edc490da29b6673f25d2fa54d2e&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Morpholine Method) min. 95.0 %