Pivalamide ≥98.0% (by GC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

P0819-25G P0819-500G P0819-100G
TCIAP0819-25GEA 58.3 EUR
TCIAP0819-25G TCIAP0819-500G TCIAP0819-100G
Pivalamide ≥98.0% (by GC, total nitrogen)
Pivalamide
Formule: C₅H₁₁NO
Kookpunt: 212°C
Dichtheid: 1,02 g/cm³ (20 °C)
MDL Nummer: MFCD00008011
CAS nummer: 754-10-9
EINECS: 212-043-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57a20f6ca7944adc3bcd88f95b204338c6f33dc4&vid=57a20f6ca7944adc3bcd88f95b204338c6f33dc4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 153.0 to 156.0 deg-C
Solubility in Methanol within almost transparenc


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR