Ethyl-4-bromobenzoate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
4-Bromobenzoic acid ethyl ester, Ethyl 4-bromobenzoate
B1515-25G
B1515-250G
TCIAB1515-25GEA
45.1
EUR
TCIAB1515-25G
TCIAB1515-250G
Ethyl-4-bromobenzoate ≥98.0% (by GC)
Ethyl-4-bromobenzoate
Formule:
C₉H₉BrO₂ Kookpunt: 131°C Smeltpunt: 18 °C Dichtheid: 1,4332 g/cm³ (17.3 °C) |
MDL Nummer:
MFCD00016329 CAS nummer: 5798-75-4 EINECS: 227-347-2 Merck Index: 14,01408 |
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...