4-Chlorobenzhydrazide ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
C2060-25G C2060-5G
TCIAC2060-25GEA 181 EUR
TCIAC2060-25G TCIAC2060-5G
4-Chlorobenzhydrazide ≥98.0% (by GC, titration analysis)
4-Chlorobenzhydrazide
Formule: C₇H₇ClN₂O
Kookpunt: 354 °C (1013 hPa)
Smeltpunt: 164 °C
MDL Nummer: MFCD00007603
CAS nummer: 536-40-3
EINECS: 208-632-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99cf0969ba02dba3c648fb13dff6d630ebb80689&vid=99cf0969ba02dba3c648fb13dff6d630ebb80689&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 163.0 to 168.0 deg-C