4-Ethoxybenzoyl chloride ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
E0700-25G
E0700-5G
TCIAE0700-25GEA
148
EUR
TCIAE0700-25G
TCIAE0700-5G
4-Ethoxybenzoyl chloride ≥98.0% (by GC, titration analysis)
4-Ethoxybenzoyl chloride
Formule:
C₉H₉ClO₂ Kookpunt: 145°C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00000688 CAS nummer: 16331-46-7 UN: 3261 ADR: 8,II |
|
Specification Test Results
Appearance | Colorless to Light yellow to Light red clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...