4-Anisoyl chloride ≥99.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
4-Methoxybenzoyl chloride
M0721-25G
M0721-500G
M0721-100G
TCIAM0721-25GEA
25.3
EUR
TCIAM0721-25G
TCIAM0721-500G
TCIAM0721-100G
4-Anisoyl chloride ≥99.0% (by GC)
4-Anisoyl chloride
Formule:
C₈H₇ClO₂ MW: 170.6 g/mol Kookpunt: 263 Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00000687 CAS nummer: 100-07-2 EINECS: 202-816-4 UN: 1729 ADR: 8 II Merck Index: 14,00673 |
|
Specification Test Results
Appearance | White or Colorless to Light orange to Yellow powder to lump to clear liquid |
Purity (GC) | min. 99.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...