Benzanilide ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

B0016-25G B0016-500G
TCIAB0016-25GEA 17.6 EUR
TCIAB0016-25G TCIAB0016-500G
Benzanilide ≥98.0% (by GC)
Benzanilide
Formule: C₁₃H₁₁NO
Kookpunt: 247 °C (1013 hPa)
Smeltpunt: 162…165 °C
Dichtheid: 1,32 g/cm³ (20 °C)
MDL Nummer: MFCD00003069
CAS nummer: 93-98-1
EINECS: 202-292-7
Merck Index: 14,01061
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f085b8f9a1a850359e17350427eda6b19df3dee2&vid=f085b8f9a1a850359e17350427eda6b19df3dee2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 163.0 to 167.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR