m,α,α,α-Tetrafluoroanisole ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

F0569-25G F0569-5G
TCIAF0569-25GEA 514 EUR
TCIAF0569-25G TCIAF0569-5G
m,α,α,α-Tetrafluoroanisole ≥98.0% (by GC)
m,α,α,α-Tetrafluoroanisole
Formule: C₇H₄F₄O
Kookpunt: 105°C
MDL Nummer: MFCD00236323
CAS nummer: 1077-01-6
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d56a3a36a7ffa1bec329b12d07e2ac9e47cc72d8&vid=d56a3a36a7ffa1bec329b12d07e2ac9e47cc72d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR