4-(Trifluoromethoxy)phenol ≥95.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: α,α,α-Trifluoro-4-hydroxyanisole
T1341-25G T1341-5G
TCIAT1341-25GEA 283 EUR
TCIAT1341-25G TCIAT1341-5G
4-(Trifluoromethoxy)phenol ≥95.0% (by GC)
4-(Trifluoromethoxy)phenol
Formule: C₇H₅F₃O₂
MW: 178.11 g/mol
Kookpunt: 95°C
Vlampunt: 86°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00040988
CAS nummer: 828-27-3
EINECS: 212-583-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a80e6ebfd98dfa52774cbfe44a41c1db72802042&vid=a80e6ebfd98dfa52774cbfe44a41c1db72802042&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 95.0 %