3-Fluorobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
F0790-25G F0790-5G
TCIAF0790-25GEA 69.3 EUR
TCIAF0790-25G TCIAF0790-5G
3-Fluorobenzenesulphonyl chloride ≥98.0% (by GC, titration analysis)
3-Fluorobenzenesulphonyl chloride
Formule: C₆H₄ClFO₂S
Kookpunt: 113°C
Smeltpunt: 7°C
MDL Nummer: MFCD00042286
CAS nummer: 701-27-9
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7535df969eda0d819bb5e1d7e98382ab7ce09ba&vid=c7535df969eda0d819bb5e1d7e98382ab7ce09ba&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %