Benzenesulphonohydrazide ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: Benzenesulphonic acid hydrazide, Benzenesulphonyl hydrazide

B0038-25G B0038-500G B0038-100G
TCIAB0038-25GEA 34.1 EUR
TCIAB0038-25G TCIAB0038-500G TCIAB0038-100G
Benzenesulphonohydrazide ≥98.0% (by titrimetric analysis)
Benzenesulphonohydrazide
Formule: C₆H₈N₂O₂S
MW: 172.21 g/mol
Smeltpunt: 200
MDL Nummer: MFCD00007583
CAS nummer: 80-17-1
EINECS: 201-255-2
UN: 3226
ADR: 4.1
Merck Index: 14,07598
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=818c5a7e5a9901ed7adb8fc838e89253880acd4b&vid=818c5a7e5a9901ed7adb8fc838e89253880acd4b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Bromination Method) min. 98.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR