4-Ethynylaniline ≥98.0% (by HPLC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
E0505-10G
E0505-25G
TCIAE0505-10GEA
192
EUR
TCIAE0505-10G
TCIAE0505-25G
4-Ethynylaniline ≥98.0% (by HPLC)
4-Ethynylaniline
Formule:
C₈H₇N |
MDL Nummer:
MFCD00168824 CAS nummer: 14235-81-5 |
|
Specification Test Results
Appearance | Light yellow to Brown to Dark green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 93.0 % |
Melting point | 102.0 to 105.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...