2-Bromocumene ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
1-Bromo-2-isopropylbenzene
B2044-25G
B2044-5G
TCIAB2044-25GEA
84.7
EUR
TCIAB2044-25G
TCIAB2044-5G
2-Bromocumene ≥97.0% (by GC)
2-Bromocumene
Formule:
C₉H₁₁Br Kookpunt: 210°C |
MDL Nummer:
MFCD00051567 CAS nummer: 7073-94-1 EINECS: 230-370-0 ADR: 9,III |
|
Specification Test Results
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...