3-Bromocumene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 1-Bromo-3-isopropylbenzene

B2875-25G B2875-5G
TCIAB2875-25GEA 127 EUR
TCIAB2875-25G TCIAB2875-5G
3-Bromocumene ≥98.0% (by GC)
3-Bromocumene
Formule: C₉H₁₁Br
Kookpunt: 210°C
MDL Nummer: MFCD01318112
CAS nummer: 5433-01-2
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f7a16e5a88e59dbec91e8086f0b3fcd73238d118&vid=f7a16e5a88e59dbec91e8086f0b3fcd73238d118&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR