4-Bromocumene ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
4-Bromoisopropylbenzene, 1-Bromo-4-isopropylbenzene
B2045-25G
B2045-5G
TCIAB2045-25GEA
161
EUR
TCIAB2045-25G
TCIAB2045-5G
4-Bromocumene ≥97.0% (by GC)
4-Bromocumene
Formule:
C₉H₁₁Br MW: 199.09 g/mol Kookpunt: 219°C Smeltpunt: –22°C Vlampunt: 94°C |
MDL Nummer:
MFCD00039159 CAS nummer: 586-61-8 EINECS: 209-577-5 |
|
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...