α-Bromo-3-chloro-4-fluorotoluene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

C2482-25G C2482-5G
TCIAC2482-25GEA 240 EUR
TCIAC2482-25G TCIAC2482-5G
α-Bromo-3-chloro-4-fluorotoluene ≥98.0% (by GC)
α-Bromo-3-chloro-4-fluorotoluene
Formule: C₇H₅BrClF
Kookpunt: 65°C
Smeltpunt: 37 °C
MDL Nummer: MFCD01631551
CAS nummer: 192702-01-5
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=359857f51460df37facbc0e186bd80f6c48c4645&vid=359857f51460df37facbc0e186bd80f6c48c4645&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR