ω-Bromo-4-fluoroacetophenone ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Bromomethyl p-fluorophenyl ketone, α-Bromo-4-fluoroacetophenone, 2-Bromo-1-(4-fluorophenyl)ethan-1-one, 2-Bromo-4'-fluoroacetophenone, 4-Fluorophenacyl bromide, 2-Bromo-1-(4-fluorophenyl)ethanone
B2382-25G
B2382-5G
TCIAB2382-25GEA
130
EUR
TCIAB2382-25G
TCIAB2382-5G
ω-Bromo-4-fluoroacetophenone ≥97.0% (by GC)
ω-Bromo-4-fluoroacetophenone
Formule:
C₈H₆BrFO Kookpunt: 153°C Smeltpunt: 49 °C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00040830 CAS nummer: 403-29-2 EINECS: 206-955-1 UN: 3261 ADR: 8,III |
|
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 46.0 to 50.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...