4-Chloro-3-nitrobenzyl alcohol ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C1712-25G
C1712-5G
TCIAC1712-25GEA
186
EUR
TCIAC1712-25G
TCIAC1712-5G
4-Chloro-3-nitrobenzyl alcohol ≥98.0% (by GC)
4-Chloro-3-nitrobenzyl alcohol
Formule:
C₇H₆ClNO₃ Smeltpunt: 66 °C |
MDL Nummer:
MFCD00007086 CAS nummer: 55912-20-4 EINECS: 259-901-4 |
|
Specification Test Results
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 63.0 to 67.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...